Allsigns International™ 1mm Rigid Plastic Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: SA27/R.

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12359817

Quantity Price
1 £ 14.870000000000001 / Each
Estimated Shipment
08-05-2017
Add to basket


Description and Specification

Specification

Certifications/Compliance EEC 92/58/EEC
Quantity 1Each
Dimensions (L x W) 300 x 150mm
Material Rigid Plastic
Message Fire exit with right running man
Type Safety sign
Documentation

Technical documents