Allsigns International™ 1mm Rigid Plastic Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: SA34R

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12359807

Quantity Price
1 £ 13.01 / Each
Estimated Shipment
08-05-2017
Add to basket


Description and Specification

Specification

Material Rigid Plastic
Type Safety sign
Quantity 1Each
Certifications/Compliance EEC 92/58/EEC
Dimensions (L x W) 200 x 200mm
Message Fire exit with down left pointing arrow
Documentation

Technical documents