missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Regulation Safety Sign

Brand:  Allsigns International™ IRR13R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12345568

  • POA / Each


Estimated Shipment
04-08-2017
Add to basket

Specifications Description & Specifications

Specifications

IRR99
1mm rigid plastic
1Each
100 x 75mm
Radioactive sources - store
Regulation Safety Sign
Documentation

Technical Documents