missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Regulation Safety Sign

Brand:  Allsigns International™ IRR2R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12325548

  • POA / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Description

Regulation sign radiation controlled area authorised persons only 200mm x 300mm 1mm rigid plastic Allsigns
Description & Specifications

Specifications

Rad. control area - x-rays fixed no empty red light
1Each
IRR99
200 x 300mm
1mm rigid plastic
Regulation Safety Sign
Documentation

Technical Documents