Allsigns International™ 1mm Rigid Plastic Prohibition Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: PR69R

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345738

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
12-06-2017
Add to basket


Description and Specification

Specification

Material Rigid Plastic
Dimensions (L x W) 200 x 300mm
Message No Exit
Color Red circle and band, black symbol
Type Prohibition Safety Sign
Quantity 1Each
Documentation

Technical documents