missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Multipurpose Safety Sign

Brand:  Allsigns International™ CO5R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12305538

  • POA / Each


Estimated Shipment
07-08-2017
Add to basket

Specifications Description & Specifications

Specifications

Danger hazardous/Protective
400 x 300mm
1Each
1mm Rigid Plastic
Multi Purpose Safety Sign
Documentation

Technical Documents