missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Multipurpose Safety Sign

Brand:  Allsigns International™ MU6R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12305528

  • POA / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

1Each
LPG highly flammable/no smoking
600 x 450mm
1mm rigid plastic
Multipurpose Safety Sign
Documentation

Technical Documents