missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Multipurpose Safety Sign

Brand:  Allsigns International™ MU9R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12345528

  • POA / Each


Estimated Shipment
15-06-2017
Add to basket

Specifications Description & Specifications

Specifications

1mm rigid plastic
600 x 450mm
1Each
Multipurpose Safety Sign
Highly flammable/no smoking
Documentation

Technical Documents