Allsigns International™ 1mm Rigid Plastic Multipurpose Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: CO10/PVC.

Safety sign CO10/PVC multi purpose sign permit towork must be obtained 400mm x 300mm 1mm rigid

UNSPSC: 55121700

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345538

Quantity Price
1 £ 24.89 / Each
Estimated Shipment
02-03-2017
Add to basket


Description and Specification

Specification

Material 1mm Rigid Plastic
Message Hazardous area/permit to work
Quantity 1Each
Type Multi Purpose Safety Sign
Dimensions (L x W) 400 x 300mm
Documentation

Technical documents