Allsigns International™ 1mm Rigid Plastic Mandatory Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: MA110/PVC.

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12349987

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
07-04-2017
Add to basket


Description and Specification

Specification

Message Safety helmets must be worn in this area
Type Mandatory Safety Sign
Quantity 1Each
Dimensions (L x W) 200 x 300mm
For Use With (Application) These signs are designed to be used to indicate suitable Exit routes and Safe areas for disabled people
Material 1mm Rigid Plastic
Color Blue symbol and background white text
Documentation

Technical documents