Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA64/PVC.

Safety sign WA64/PVC hazard warning cautionfork-lift trucks operating 200mm x 300mm 1mm

UNSPSC: 55121700

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12305628

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
24-02-2017
Add to basket


Description and Specification

Specification

Material 1mm Rigid Plastic
Message Caution fork lift trucks operating
Quantity 1Each
Type Hazard Warning Safety Sign
Dimensions (L x W) 200 x 300mm