missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign

Brand:  Allsigns International™ WA32R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12385608

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Specifications

Material: Rigid Plastic, 1mm
Danger buried cables
300mm
1/Ea.
200mm
Hazard Warning Safety Sign
Plastic
Documentation

Technical Documents