missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign

Brand:  Allsigns International™ WA7R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12335598

  • POA / Each


Estimated Shipment
02-08-2017
Add to basket

Specifications Description & Specifications

Specifications

Hazard Warning Safety Sign
1Each
1mm Rigid Plastic
Danger 240 volts
200 x 300mm
Documentation

Technical Documents