Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA123/PVC.

Safety sign WA123/PVC hazard warning caution veryhot water 75mm x 50mm 1mm rigid plastic Allsigns,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385648

Quantity Price
1 £ 9.66 / Each
Estimated Shipment
02-03-2017
Add to basket


Description and Specification

Specification

Material 1mm Rigid Plastic
Message Caution Very Hot Water
Quantity 1Each
Type Hazard Warning Safety Sign
Dimensions (L x W) 75 x 50mm