Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA148/PVC.

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12325678

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
16-03-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
Material 1mm rigid plastic
Type Hazard Warning Safety Sign
Message Spontaneously combustible
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents