Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: EC33R

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345708

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
11-05-2017
Add to basket


Description and Specification

Specification

Type Hazard Warning Safety Sign
Message Risk of Freezing
Quantity 1Each
Dimensions (L x W) 200 x 300mm
Material 1mm Rigid Plastic
Documentation

Technical documents