Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA149R

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345678

Quantity Price
1 £ 19.83 / Each
Estimated Shipment
07-06-2017
Add to basket


Description and Specification

Specification

Message Organic Peroxide
Color Black Symbol and Text, Yellow Background
Type Hazard Warning Safety Sign
Dimensions (L x W) 200 x 300mm
For Use With (Application) To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace
Material 1mm Rigid Plastic
Quantity 1Each

To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace.Colour - black symbol and text, yellow backgroundSymbol - triangleS/A - Self-Adhesive

Documentation