missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign

Brand:  Allsigns International™ WA66R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12325628

  • POA / Each


Estimated Shipment
07-08-2017
Add to basket

Specifications Description & Specifications

Specifications

LPG highly flammable
Hazard Warning Safety Sign
1mm rigid plastic
1Each
200 x 300mm