Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA140R

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385658

Quantity Price
1 £ 21.22 / Each
Estimated Shipment
09-05-2017
Add to basket


Description and Specification

Specification

Material 1mm Rigid Plastic
Dimensions (L x W) 200 x 300mm
Quantity 1Each
Type Hazard Warning Safety Sign
Message Harmful Substance
Documentation

Technical documents