Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA83R

UNSPSC: 55121700

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385628

Quantity Price
1 £ 20.85 / Each
Estimated Shipment
17-05-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
Material 1mm Rigid Plastic
Message Caution trip hazard
Dimensions (L x W) 200 x 300mm
Type Hazard Warning Safety Sign