Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA146R

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385668

Quantity Price
1 £ 21.94 / Each
Estimated Shipment
07-04-2017
Add to basket


Description and Specification

Specification

Material 1mm rigid plastic
Quantity 1Each
Dimensions (L x W) 200 x 300mm
Message Flammable gas
Color Black symbol and text, Yellow background
Type Hazard Warning Safety Sign
Adhesive Type Self-adhesive

To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace.Colour - black symbol and text, yellow backgroundSymbol - triangleS/A - Self-Adhesive

Documentation