Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA147/PVC.

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12305678

Call for latest availability
+44 (0)1509 555500
Add to basket


Description and Specification

Specification

Type Hazard Warning Safety Sign
Material 1mm Rigid Plastic
Quantity 1Each
Message Dangerous When Wet
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents