Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA26A/PVC.

Safety sign WA26A/PVC hazard warning danger ofdeath 200mm x 300mm 1mm rigid plastic Allsigns,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345608

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
31-03-2017
Add to basket


Description and Specification

Specification

Material 1mm Rigid Plastic
Message Danger of death
Quantity 1Each
Type Hazard Warning Safety Sign
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents