Allsigns International™ 1mm Rigid Plastic Hazard Warning Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA15/PVC.

Safety sign WA15/PVC hazard warning danger livewires 200mm x 300mm 1mm rigid plastic Allsigns,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12325608

Quantity Price
1 £ 21.79 / Each
Estimated Shipment
03-04-2017
Add to basket


Description and Specification

Specification

Material 1mm Rigid Plastic
Message Danger live wires
Quantity 1Each
Type Hazard Warning Safety Sign
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents