Allsigns International™ 1mm Rigid Plastic Condition Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA215/PVC.

Safety sign WA215/PVC hazard warning automaticdoor arrow pointing left 200mm x 75mm 1mm rigid

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12325648

Quantity Price
1 £ 9.38 / Each
Estimated Shipment
30-03-2017
Add to basket


Description and Specification

Specification

Material Rigid Plastic
Message Automatic door left
Quantity 1Each
Type Hazard Warning Safety Sign
Dimensions (L x W) 200 x 75mm