Allsigns International™ 1mm Rigid Plastic Condition Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: SA81R

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12349907

Quantity Price
1 £ 12.21 / Each
Estimated Shipment
09-06-2017
Add to basket


Description and Specification

Specification

Material Rigid Plastic
Dimensions (L x W) 300 x 150mm
Message Push bar to open
Quantity 1Each
Color White symbol and text, Green background
Type Condition Safety Sign
Documentation

Technical documents