missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ 1mm Rigid Plastic Condition Safety Sign

Brand:  Allsigns International™ SA93R

UNSPSC : 55121712

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12389917

  • POA / Each


Estimated Shipment
07-08-2017
Add to basket

Specifications Description & Specifications

Specifications

White symbol and text, Green background
Rigid Plastic
1Each
75 x 150mm
Pull
Condition Safety Sign
Documentation

Technical Documents