Allsigns International™ 1mm Rigid Plastic Condition Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA216R

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12345648

Quantity Price
1 £ 9.38 / Each
Estimated Shipment
09-06-2017
Add to basket


Description and Specification

Specification

Type Hazard Warning Safety Sign
Message Caution glass door
Material Rigid Plastic
Dimensions (L x W) 200 x 75mm
Quantity 1Each