missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrrole-2-carbohydrazide, 97%, Thermo Scientific™

Catalog Number p-2695242
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
50269-95-9
C5H7N3O
125.131
MFCD00141885
FQIAWPZQJCAPSW-UHFFFAOYSA-N
pyrrolecarbohydrazide, 2-pyrrolecarbohydrazide, pyrrole-2-carbohydrazide, pyrrole-5-carboxhydrazide, 2-pyrrolecarboxylic acid hydrazide, 2-pyrrolecarboxylic acid, hydrazide, pyrrole-2-carboxylic acid hydrazide, 1h-pyrrole-2-carboxylic acid hydrazide, 1h-pyrrole-2-carboxylicacid, hydrazide, 1h-pyrrole-2-carboxylic acid, hydrazide 9ci
281610
1H-pyrrole-2-carbohydrazide
C1=CNC(=C1)C(=O)NN
This item is not returnable. View return policy
50269-95-9
C5H7N3O
125.131
MFCD00141885
FQIAWPZQJCAPSW-UHFFFAOYSA-N
pyrrolecarbohydrazide, 2-pyrrolecarbohydrazide, pyrrole-2-carbohydrazide, pyrrole-5-carboxhydrazide, 2-pyrrolecarboxylic acid hydrazide, 2-pyrrolecarboxylic acid, hydrazide, pyrrole-2-carboxylic acid hydrazide, 1h-pyrrole-2-carboxylic acid hydrazide, 1h-pyrrole-2-carboxylicacid, hydrazide, 1h-pyrrole-2-carboxylic acid, hydrazide 9ci
281610
1H-pyrrole-2-carbohydrazide
C1=CNC(=C1)C(=O)NN
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 1H-pyrrole-2-carbohydrazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'