missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Pyrazole-1-carboxamidine monohydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 167°C to 170°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (Argentometry) |
Packaging | Glass bottle |
Quantity | 10 g |
Solubility Information | Solubility in water: soluble |
Formula Weight | 146.58 |
Show More |
Safety and Handling
Product Identifier
- 1H-Pyrazole-1-carboxamidine monohydrochloride
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'