missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-3-carbonitrile, 97%, Maybridge™

Catalog Number p-2695327
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
5457-28-3
C9H6N2
142.16
MFCD00022717
CHIFTAQVXHNVRW-UHFFFAOYSA-N
3-cyanoindole, indole-3-carbonitrile, 3-indolecarbonitrile, chembl46724, zlchem 360, 3-cyano-1h-indole, pubchem7325, acmc-209lhi, 1h-indol-3-yl cyanide
230282
1H-indole-3-carbonitrile
N#CC1=CNC2=CC=CC=C12
This item is not returnable. View return policy
5457-28-3
C9H6N2
142.16
MFCD00022717
CHIFTAQVXHNVRW-UHFFFAOYSA-N
3-cyanoindole, indole-3-carbonitrile, 3-indolecarbonitrile, chembl46724, zlchem 360, 3-cyano-1h-indole, pubchem7325, acmc-209lhi, 1h-indol-3-yl cyanide
230282
1H-indole-3-carbonitrile
N#CC1=CNC2=CC=CC=C12
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 142.16
Chemical Name or Material 1H-Indole-3-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'