missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-2-carboxylic acid, 97%

Brand:  Maybridge CC12801DE

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 1477-50-5 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 10536585

  • £8.00 / 5g


Estimated Shipment date
from Supplier 27-09-2017
Add to basket

Specifications

Chemical Identifiers

1H-Indole-2-carboxylic acid
97%
161.16
97%
C9H7NO2

Specifications

1H-Indole-2-carboxylic acid
97%
C9H7NO2
5g
1477-50-5
97%
161.16
Documentation

Technical Documents