missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Indene, 90+%, stab. with 0.01% 4-tert-butylcatechol, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Description
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.
Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.
Notes
Store at 4°C. Store in dark. Incompatible with oxidizing agent, light, heat.
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.
Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.
Notes
Store at 4°C. Store in dark. Incompatible with oxidizing agent, light, heat.
Specifications
Melting Point | ∽-2°C |
Density | 0.996 |
Boiling Point | 181°C to 182°C |
Flash Point | 58°C (136°F) |
Assay Percent Range | ≥90% |
Refractive Index | 1.57 |
Quantity | 50 mL |
Beilstein | 635873 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in ethanol,acetone,ethyl ether,and ether. Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'