missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole-7-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10574248
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
677304-69-7
C8H6N2O2
162.15
MFCD06804572
WBCWIQCXHSXMDH-UHFFFAOYSA-N
7-carboxy-1h-indazole, 1h-indazole-7-carboxylicacid, indazole-7-carboxylic acid, 7-indazole carboxylic acid, pubchem17673, acmc-1bl1h, ksc351o8t, 2h-indazole-7-carboxylic acid, 7-1h indazole carboxylic acid
12639205
1H-indazole-7-carboxylic acid
C1=CC2=C(C(=C1)C(=O)O)NN=C2
This item is not returnable. View return policy
677304-69-7
C8H6N2O2
162.15
MFCD06804572
WBCWIQCXHSXMDH-UHFFFAOYSA-N
7-carboxy-1h-indazole, 1h-indazole-7-carboxylicacid, indazole-7-carboxylic acid, 7-indazole carboxylic acid, pubchem17673, acmc-1bl1h, ksc351o8t, 2h-indazole-7-carboxylic acid, 7-1h indazole carboxylic acid
12639205
1H-indazole-7-carboxylic acid
C1=CC2=C(C(=C1)C(=O)O)NN=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 229.0°C to 231.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 162.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazole-7-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Indazole-7-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'