missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-indazole-3-carboxylic acid, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 162.15 |
Percent Purity | 97% |
Chemical Name or Material | 1H-Indazole-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'