missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10683225
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1072-84-0
C4H4N2O2
112.09
MFCD00082203
NKWCGTOZTHZDHB-UHFFFAOYSA-N
1h-imidazole-4-carboxylic acid, 4-imidazolecarboxylic acid, imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4-carboxyimidazole, unii-ae82z8u4gj, imidazole-5-carboxylic acid, 4-imidazole carboxylic acid, ae82z8u4gj, 4 5-imidazolecarboxylic acid
14080
1H-imidazole-5-carboxylic acid
OC(=O)C1=CN=CN1
This item is not returnable. View return policy
1072-84-0
C4H4N2O2
112.09
MFCD00082203
NKWCGTOZTHZDHB-UHFFFAOYSA-N
1h-imidazole-4-carboxylic acid, 4-imidazolecarboxylic acid, imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4-carboxyimidazole, unii-ae82z8u4gj, imidazole-5-carboxylic acid, 4-imidazole carboxylic acid, ae82z8u4gj, 4 5-imidazolecarboxylic acid
14080
1H-imidazole-5-carboxylic acid
OC(=O)C1=CN=CN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 242°C to 244°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 112.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1H-Imidazole-4-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Imidazole-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'