missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluoro-1-tetradecanol, 96%, Thermo Scientific Chemicals

Catalog Number p-7046865
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
15622-57-8
C14H3F27O
700.134
MFCD00153264
WYCXYEWKMLSDBQ-UHFFFAOYSA-N
1h,1h-perfluoro-1-tetradecanol, heptacosafluorotridecyl methanol, 1h,1h-perfluorotetradecan-1-ol
2776373
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecan-1-ol
C(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
This item is not returnable. View return policy
15622-57-8
C14H3F27O
700.134
MFCD00153264
WYCXYEWKMLSDBQ-UHFFFAOYSA-N
1h,1h-perfluoro-1-tetradecanol, heptacosafluorotridecyl methanol, 1h,1h-perfluorotetradecan-1-ol
2776373
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecan-1-ol
C(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 137°C to 140°C
Boiling Point 130°C (60 mmHg)
Quantity 5 g
Formula Weight 700.14
Percent Purity 96%
Chemical Name or Material 1H,1H-Perfluoro-1-tetradecanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'