missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1H,1H,8H,8H-Dodecafluoro-1,8-octanediol, 97%

£16.20 - £40.40

Chemical Identifiers

CAS 90177-96-1
Molecular Formula C8H6F12O2
Molecular Weight (g/mol) 362.115
MDL Number MFCD00153145
InChI Key XZJPYETUABEQFI-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-perfluorooctane-1,8-diol, 1,8-dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane, 1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, 1h,1h,8h,8h-perfluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluorooctane-1,8-diol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octane-1,8-diol
PubChem CID 3820533
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol
SMILES C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11319195 SDS View Documents Alfa Aesar™
L16484.03
1g £16.20
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-11-2019
Please to purchase this item. Need a web account? Register with us today!
11329195 SDS View Documents Alfa Aesar™
L16484.06
5g £40.40
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-11-2019
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
1H,1H,8H,8H-Dodecafluoro-1,8-octanediol is used in pharma industry.

Solubility
Soluble in methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

90177-96-1
362.115
XZJPYETUABEQFI-UHFFFAOYSA-N
3820533
C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
C8H6F12O2
MFCD00153145
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-perfluorooctane-1,8-diol, 1,8-dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane, 1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, 1h,1h,8h,8h-perfluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluorooctane-1,8-diol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octane-1,8-diol
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol

Specifications

1g
105°C (0.08mmHg)
90177-96-1
HOCH2(CF2)6CH2OH
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-perfluorooctane-1,8-diol, 1,8-dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane, 1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, 1h,1h,8h,8h-perfluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluorooctane-1,8-diol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octane-1,8-diol
C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
362.115
362.12
1H,1H,8H,8H-Dodecafluoro-1,8-octanediol
80°C to 88°C
C8H6F12O2
MFCD00153145
XZJPYETUABEQFI-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol
3820533
97%