missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,7H-Dodecafluoro-1-heptanol, 97%, Thermo Scientific Chemicals

Catalog Number 11420681
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
335-99-9
C7H4F12O
332.089
MFCD00039630
BYKNGMLDSIEFFG-UHFFFAOYSA-N
1h,1h,7h-dodecafluoro-1-heptanol, 1,1,7-trihydroperfluoroheptanol, 1h,1h,7h-dodecafluoroheptanol, 1-heptanol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, unii-lpx5k7ec3w, 1,1,7-trihydrododecafluoroheptanol, 1,1,7-trihydroperfluoroheptyl alcohol, 1-heptanol, 1h,1h,7h-dodecafluoro, 1,1,7-trihydrododecafluoroheptan-1-ol, 1h,1h,7h-dodecafluoro-1-hydroxyheptane
67639
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptan-1-ol
C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
This item is not returnable. View return policy
335-99-9
C7H4F12O
332.089
MFCD00039630
BYKNGMLDSIEFFG-UHFFFAOYSA-N
1h,1h,7h-dodecafluoro-1-heptanol, 1,1,7-trihydroperfluoroheptanol, 1h,1h,7h-dodecafluoroheptanol, 1-heptanol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, unii-lpx5k7ec3w, 1,1,7-trihydrododecafluoroheptanol, 1,1,7-trihydroperfluoroheptyl alcohol, 1-heptanol, 1h,1h,7h-dodecafluoro, 1,1,7-trihydrododecafluoroheptan-1-ol, 1h,1h,7h-dodecafluoro-1-hydroxyheptane
67639
2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptan-1-ol
C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -20°C
Density 1.762
Boiling Point 169°C to 170°C
Flash Point None
Refractive Index 1.318
Quantity 25 g
Beilstein 1800110
Formula Weight 332.09
Percent Purity 97%
Chemical Name or Material 1H,1H,7H-Dodecafluoro-1-heptanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'