missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluoro-1-dodecanol, 96%, Thermo Scientific™

Catalog Number p-7058456
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
865-86-1
C12H5F21O
564.14
MFCD00039545
FLXYIZWPNQYPIT-UHFFFAOYSA-N
2-perfluorodecyl ethanol, 1h,1h,2h,2h-perfluorododecan-1-ol, 1,1,2,2-tetrahydroperfluoro dodecanol, 1h,1h,2h,2h-perfluoro-1-dodecanol, unii-a72u7q973t, 1h,1h,2h,2h-perfluorododecanol, 1,1,2,2-tetrahydroperfluoro-1-dodecanol, 1h,1h,2h,2h-perfluorodeca-1-ol, 1h,1h,2h,2h-perfluoro-1-dodecanol 1g
70083
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecan-1-ol
OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
This item is not returnable. View return policy
865-86-1
C12H5F21O
564.14
MFCD00039545
FLXYIZWPNQYPIT-UHFFFAOYSA-N
2-perfluorodecyl ethanol, 1h,1h,2h,2h-perfluorododecan-1-ol, 1,1,2,2-tetrahydroperfluoro dodecanol, 1h,1h,2h,2h-perfluoro-1-dodecanol, unii-a72u7q973t, 1h,1h,2h,2h-perfluorododecanol, 1,1,2,2-tetrahydroperfluoro-1-dodecanol, 1h,1h,2h,2h-perfluorodeca-1-ol, 1h,1h,2h,2h-perfluoro-1-dodecanol 1g
70083
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecan-1-ol
OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluoro-1-dodecanol is used in fabric processing, coating, appliance paint and boat paint.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 90°C to 95°C
Boiling Point 145°C (10 mmHg)
Linear Formula CF3(CF2)9CH2CH2OH
Quantity 1 g
Beilstein 2029867
Solubility Information Insoluble in water.
Formula Weight 564.14
Percent Purity 96%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-dodecanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'