missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Perfluoro-1-dodecanol, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1H,1H,2H,2H-Perfluoro-1-dodecanol is used in fabric processing, coating, appliance paint and boat paint.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
1H,1H,2H,2H-Perfluoro-1-dodecanol is used in fabric processing, coating, appliance paint and boat paint.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
Specifications
Melting Point | 90°C to 95°C |
Boiling Point | 145°C (10 mmHg) |
Linear Formula | CF3(CF2)9CH2CH2OH |
Quantity | 5 g |
Beilstein | 2029867 |
Solubility Information | Insoluble in water. |
Formula Weight | 564.14 |
Percent Purity | 96% |
Chemical Name or Material | 1H,1H,2H,2H-Perfluoro-1-dodecanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'