missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1H,1H,11H-Perfluoro-1-undecanol, tech. 85%

£12.08 - £62.43

Chemical Identifiers

CAS 307-70-0
Molecular Formula C11H4F20O
Molecular Weight (g/mol) 532.12
MDL Number MFCD00039628
InChI Key GJYLSFCFHFUODO-UHFFFAOYSA-N
Synonym 1h,1h,11h-eicosafluoro-1-undecanol, 1-undecanol, 1h,1h,11h-eicosafluoro, 1,1,11-trihydroperfluoroundecanol, 1h,1h,11h-perfluoro-1-undecanol, 1h,1h,11h-eicosafluoroundecanol-1, 1h,1h,11h-eicosafluoro-1-undecanol, tech., 1h,11h-eicosafluoro-1-undecanol, 1h,1h,11h-perfluoroundecan-1-ol, 1-undecanol,1h,11h-eicosafluoro, 1h, 1h, 11h-eicosafluoroundecanol
PubChem CID 67548
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecan-1-ol
SMILES C(C(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11484117 SDS View Documents Alfa Aesar™
A19552.06
5g £12.08
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-11-2019
Please to purchase this item. Need a web account? Register with us today!
11494117 SDS View Documents Alfa Aesar™
A19552.14
25g £22.15
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-11-2019
Please to purchase this item. Need a web account? Register with us today!
11494170 SDS View Documents Alfa Aesar™
A19552.22
100g £62.43
100g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-11-2019
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

307-70-0
532.12
GJYLSFCFHFUODO-UHFFFAOYSA-N
67548
C(C(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
C11H4F20O
MFCD00039628
1h,1h,11h-eicosafluoro-1-undecanol, 1-undecanol, 1h,1h,11h-eicosafluoro, 1,1,11-trihydroperfluoroundecanol, 1h,1h,11h-perfluoro-1-undecanol, 1h,1h,11h-eicosafluoroundecanol-1, 1h,1h,11h-eicosafluoro-1-undecanol, tech., 1h,11h-eicosafluoro-1-undecanol, 1h,1h,11h-perfluoroundecan-1-ol, 1-undecanol,1h,11h-eicosafluoro, 1h, 1h, 11h-eicosafluoroundecanol
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecan-1-ol

Specifications

5g
180°C to 181°C (200mmHg)
307-70-0
MFCD00039628
GJYLSFCFHFUODO-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecan-1-ol
67548
85%
1H,1H,11H-Perfluoro-1-undecanol
90°C to 95°C
C11H4F20O
1h,1h,11h-eicosafluoro-1-undecanol, 1-undecanol, 1h,1h,11h-eicosafluoro, 1,1,11-trihydroperfluoroundecanol, 1h,1h,11h-perfluoro-1-undecanol, 1h,1h,11h-eicosafluoroundecanol-1, 1h,1h,11h-eicosafluoro-1-undecanol, tech., 1h,11h-eicosafluoro-1-undecanol, 1h,1h,11h-perfluoroundecan-1-ol, 1-undecanol,1h,11h-eicosafluoro, 1h, 1h, 11h-eicosafluoroundecanol
C(C(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
532.12
532.12
Technical