missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,4-Triazole-3-carboxylic acid, 97%, ACROS Organics™

Brand:  Acros Organics 421410050

Code : 29

Additional Details : CAS Number : 4928-87-4 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11906641

  • £62.90 / 5g


Estimated Shipment
04-07-2018
Add to basket


Specifications

Chemical Identifiers

1H-1, 2, 4-Triazole-3-carboxylic acid
96.0
97%
C3H3N3O2
4928-87-4
100.0
97%
113.08

Specifications

1H-1, 2, 4-Triazole-3-carboxylic acid
96.0
Authentic
Powder
97%
113.08
White to pale brown
4928-87-4
100.0
Glass bottle
97%
C3H3N3O2
119.0° to 121.0°C
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1H-1,2,4-Triazole-3-carboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS