Learn More
Thermo Scientific™ BioBasic™ PREP SEC 60 Size Exclusion Chromatography HPLC Columns
Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 60 Size Exclusion Chromatography HPLC Columns.
Brand: Thermo Scientific™ 73305-157846A
Additional Details : Weight : 3.00000kg
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 60,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.
BioBasic™ PREP SEC 60 HPLC Column Benefits:
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 60Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
Preparative Column, Standard | |
150 mm | |
5 μm | |
60 Å | |
60°C | |
Spherical, Fully Porous, Ultrapure Silica | |
Size Exclusion | |
Size Exclusion |
7.8 mm | |
4300 psi (295 bar) | |
2 to 8 | |
450 m²/g | |
Diol (OH) | |
>9% | |
No | |
Preparative Chromatography |