missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, for HPLC, Gradient Grade, ACS, Honeywell Burdick & Jackson™
Click to view available options
Quantity:
7 L
30 L
45 L
395 L
Specifications
Melting Point | -98°C |
Color | Colorless |
CAS Max % | 1 |
Linear Formula | CH3OH |
Beilstein | 1098229 |
Grade | Far UV/Gradient |
Density | 0.79 g/cm3 (20°C) |
Boiling Point | 64°C to 65°C (1013 hPa) |
Quantity | 30 L |
Flash Point | 11°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'