missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns

Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns.

Brand:  Thermo Scientific™ 73605-157846A

Additional Details : Weight : 3.00000kg

 View more versions of this product

Product Code. 16601684

  • £1352.60 / Each
In Stock
to see stock.
Add to basket

Description

Description

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 1000 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 70,000 to 10,000,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.

BioBasic™ PREP SEC 120 HPLC Column Benefits:

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 120Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

Specifications

Specifications

Preparative Column, Standard
Preparative Size Exclusion
4300 psi (295 bar)
2 to 8
25 m²/g
L59
Spherical, Fully Porous, Ultrapure Silica
Size Exclusion
Size Exclusion
7.8 mm
150 mm
5 μm
1000 Å
60°C
Diol (OH)
>0.6%
No
Preparative Chromatography
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers