missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Twin-Strep-tag™ Capture Kit

The Twin-Strep-tag™ Capture Kit can be used for reversible capture of Twin-Strep-tag™ fusion proteins for SPR analyses

Brand:  IBA Lifesciences 2-4370-000

Additional Details : Weight : 0.02500kg

Product Code. 15989190

  • £773.00 / 1 set
Estimated Shipment: 17-04-2024
to see stock.
Add to basket

Description

Description

Twin-Strep-tag™ Capture Kit is intended for site-directed, reversible capture of Twin-Strep-tag™ fusion proteins for biomolecular interaction analysis using surface plasmon resonance (SPR) devices. The kit provides Strep-Tactin™XT, SPR Immobilization Buffer, Regeneration Buffer, and GFP-Twin-Strep-tag™ as control protein. It enables the highly stable immobilization of the ligand based on the unique Strep-Tactin™XT:Twin-Strep-tag™ interaction, which operates with pM affinity. The key benefits are:
• Indirect immobilization protecting the ligand from chemical modification as well as inactivation or manipulation by the immobilization procedure
• High affinity enabling measurements over a long period (high affinity in low pM range; T1/2 = 13 h)
• Chip can be regenerated easily by an established procedure – cost effective
• Ligands can be captured directly from complex samples due to the high affinity and specificity of the Twin-Strep-tag™:Strep-Tactin™XT interaction The use of unconjugated Strep-Tactin™XT requires a license that is provided with the purchase of the kit as limited use license.
Specifications

Specifications

>95%
Strep-Tactin™XT, 100 μL
Purification of Recombinant Fusion Proteins with HPLC or FPLC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.