Learn More
3M™ Particulate Respirator, FFP3, Valved, 8833SP
3M™ Particulate Respirator 8833 is a cup-shaped disposable respirator with a comfortable M-shaped nose clip, a soft waffle edge, and 3M™ Cool Flow™ Valve.
Brand: 3M™ 7100091323
15.80 GBP valid until 2024-03-30
Use promo code "21722" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.09000kg
Description
- Provides FFP3 (NPF 50) protection against dusts, oil-based mists and other fine particles
- Robust, cup-shaped design
- 3M™ Cool Flow™ Valve helps reduce heat build-up, even in hot and humid conditions
- Features a full-face seal for improved fit and soft, cushioned lining for long-lasting comfort
- Unique, M-shaped nose clip for extra customisation of fit
- Lightweight, twin-strap design, nose foam, and soft waffle edge helps fit comfortably to a wide range of face sizes
- Advanced Electret (filter) Media helps wearers breathe easily
- Available in large or small pack sizes
- The soft inner face-seal ring improves face seal and feeling of comfort and can be hygienically cleaned if used for longer than one shift (use 3M™ 105 Face Seal Cleaner)
- Braided headbands are coloured for easy identification: red for FFP3
Specifications
EN 149:2001+A1:2009 FFP3 R D, CE Approved | |
Respirator | |
5/Pk., 4 Pk./Cs. |