Learn More
3M™ FFP3 Unvalved Particulate Respirator Dispenser
3M™ FFP3 Unvalved Particulate Respirator Dispenser provides FFP3 breathing protection against airborne particles and non-volatile liquids.
Brand: 3M™ 7000108744
Code : NEW
Additional Details : Weight : 0.32000kg
Description
- Provides economic FFP3 (NPF 50) breathing protection against airborne particles and non-volatile liquids
- Flat-fold design makes the respirator easy to store when not in use
- Single-loop strap helps you make easy adjustments for a comfortable fit
- Diamond-style front panel helps the respirator retain shape even in hot or humid conditions
- color-coded nose clip for easy identification: red for FFP3
- Individually wrapped
- Diamond-shaped front panel allows the respirator to retain its shape even in hot or humid conditions
- Meets clogging requirements as per EN 149:2001+A1:2009, ensuring prolonged respirator life while maintaining ease of breathing
- Supplied in a handy dispenser box
- Noseclip is colored for easy identification: yellow for FFP1; blue for FFP2;
Specifications
CE Approved, EN 149: 2001 + A1: 2009 FFP3 NR D | |
White | |
Folded particle mask |