Learn More
Welch™ CRVpro™ Starter Package
The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
Brand: Welch™ 330071
Code : NEW
Additional Details : Weight : 21.00000kg
Description
- CRVpro Starter Package Includes:
- Oil mist separator AKD 16
- 2-way ball valve
- Hose connector DN 16KF-10/8 mm
- Vacuum hose (1.5 m)
- Oil change set
Specifications
CRVpro with Oil Mist Separator AKD 16, 2-Way Ball Valve, Hose Connector, Vacuum Hose (1.5 m), Oil Change Set | |
CRVpro 4 | |
Rotary Vane Pump |